Merck
  • TRIM25 Promotes TNF-α-Induced NF-κB Activation through Potentiating the K63-Linked Ubiquitination of TRAF2.
TRIM25 Promotes TNF-α-Induced NF-κB Activation through Potentiating the K63-Linked Ubiquitination of TRAF2.
Journal of immunology (Baltimore, Md. : 1950) (2020-02-07)
Yuchun Liu, Kunpeng Liu, Yingqi Huang, Meng Sun, Qingnan Tian, Shoutao Zhang, Yunfei Qin
ZUSAMMENFASSUNG
As an important effector in response to various intracellular or extracellular stimuli, the NF-κB family extensively participates in a wide spectrum of biological events, and its dysregulation may result in many pathological conditions, such as microbial infection, tumor progression, and neurodegenerative disorders. Previous investigations showed that multiple types of ubiquitination play critical roles in the modulation of the NF-κB signaling pathway, yet the molecular mechanisms are still poorly understood. In the current study, we identified TRIM25, an E3 ubiquitin ligase, as a novel positive regulator in mediating NF-κB activation in human embryonic kidney 293T (HEK293T), HeLa cells, THP-1 cells, and PBMCs. The expression of TRIM25 promoted TNF-α-induced NF-κB signaling, whereas the knockdown had the opposite effect. Furthermore, TRIM25 interacted with TRAF2 and enhanced the K63-linked polyubiquitin chains attached to TRAF2. Moreover, TRIM25 bridged the interaction of TRAF2 and TAK1 or IKKβ. To our knowledge, our study has identified a previously unrecognized role for TRIM25 in the regulation of NF-κB activation by enhancing the K63-linked ubiquitination of TRAF2.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonale ANTI-FLAG® M2-Peroxidase (HRP) in Maus hergestellte Antikörper, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-β-Actin-Antikörper, Maus monoklonal, clone AC-15, purified from hybridoma cell culture
Roche
Anti-HA-Peroxidase mit hoher Affinität, from rat IgG1
Sigma-Aldrich
IL-1 beta human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture