- Application of creatinine-sensitive biosensor for hemodialysis control.
Application of creatinine-sensitive biosensor for hemodialysis control.
Biosensors & bioelectronics (2012-03-31)
O A Zinchenko, S V Marchenko, T A Sergeyeva, A L Kukla, A S Pavlyuchenko, E K Krasyuk, A P Soldatkin, A V El'skaya
PMID22459582
ZUSAMMENFASSUNG
The highly sensitive and selective potentiometric biosensor for creatinine determination has been developed by us earlier. In it, pH-sensitive field effect transistors were used as transducer and immobilized creatinine deiminase (EC 3.5.4.21)--as a biosensitive element. In the work presented, we optimized this biosensor for creatinine analysis in real samples of dialysate in patients with renal failure. The optimized version of biosensor was applied for on-line monitoring of the level of creatinine in the patient's dialysate fluid in the course of dialysis session. High correlation between the biosensor analysis and traditional Jaffe method was demonstrated.