Merck
  • Gold nanorods with an ultrathin anti-biofouling siloxane layer for combinatorial anticancer therapy.
Gold nanorods with an ultrathin anti-biofouling siloxane layer for combinatorial anticancer therapy.
Journal of drug targeting (2020-03-03)
Jinjoo Kim, Hyeongseop Keum, Hansol Kim, Byeongjun Yu, Wonsik Jung, Changhee Whang, Changjin Seo, Ji Ho Park, Sangyong Jon
ZUSAMMENFASSUNG
Despite the wide utility of gold nanorods (GNRs) in biomedical fields, only a few methods for modifying or coating the surface of GNRs suitable for biomedical applications are available. In this study, we report a new facile method that enables formation of an ultra-thin (nanometre-thickness) siloxane layer on GNRs with anti-biofouling properties and ligand functionalisation ability. A triblock random copolymer, poly(TMSMA-r-PEGMA-r-NAS), was used to coat GNRs. An ultrathin polymeric shell was formed surrounding GNRs through acid-catalysed crosslinking of silicates of TMSMA. The polymer-coated GNRs (p-GNRs) exhibited high colloidal stability in biological solutions of high ionic strength and long-term stability superior to that of PEG2k-S-GNRs. The functionalities of NAS were demonstrated using two methods for conjugating targeting ligands and loading doxorubicin via electrostatic interactions. The ligand-specific cancer-targeting ability and combinatorial chemo-photothermal anticancer effects were validated in vitro and in vivo, suggesting their potential utility in various fields.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3-(Trimethoxysilyl)propylmethacrylat, 98%