Merck
  • Characteristics and Outcomes in Patients Presenting With COVID-19 and ST-Segment Elevation Myocardial Infarction.
Characteristics and Outcomes in Patients Presenting With COVID-19 and ST-Segment Elevation Myocardial Infarction.
The American journal of cardiology (2020-08-01)
Anas Hamadeh, Ali Aldujeli, Kasparas Briedis, Kristen M Tecson, Jorge Sanz-Sánchez, Montazar Al Dujeili, Ammar Al-Obeidi, Jose Luís Diez, Remigijus Žaliūnas, Robert C Stoler, Peter A McCullough
ZUSAMMENFASSUNG
There is limited information regarding clinical characteristics and outcomes of patients with SARS-CoV-2 (COVID-19) disease presenting with ST-segment elevation myocardial infarction (STEMI). In this multicenter retrospective study, we reviewed charts of patients admitted with symptomatic COVID-19 infection and STEMI to a total of 4 hospitals spanning Italy, Lithuania, Spain and Iraq from February 1, 2020 to April 15, 2020. A total of 78 patients were included in this study, 49 (63%) of whom were men, with a median age of 65 [58, 71] years, and high comorbidity burden. During hospitalization, 8 (10%) developed acute respiratory distress syndrome, and 14 (18%) required mechanical ventilation. 19 (24%) patients were treated with primary Percutaneous Coronary Intervention (PCI) and 59 (76%) were treated with fibrinolytic therapy. 13 (17%) patients required cardiac resuscitation, and 9 (11%) died. For the 19 patients treated with primary PCI, 8 (42%) required intubation and 8 (42%) required cardiac resuscitation; stent thrombosis occurred in 4 patients (21%). A total of 5 patients (26%) died during hospitalization. 50 (85%) of the 59 patients initially treated with fibrinolytic therapy had successful fibrinolysis. The median time to reperfusion was 27 minutes [20, 34]. Hemorrhagic stroke occurred in 5 patients (9%). Six patients (10%) required invasive mechanical ventilation; 5 (9%) required cardiac resuscitation, and 4 (7%) died. In conclusion, this is the largest case series to-date of COVID-19 positive patients presenting with STEMI and spans 4 countries. We found a high rate of stent thrombosis, indicating a possible need to adapt STEMI management for COVID-19 patients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Suplatast tosylate, ≥98% (HPLC)