Merck
  • Defects in mRNA Translation in LRRK2-Mutant hiPSC-Derived Dopaminergic Neurons Lead to Dysregulated Calcium Homeostasis.
Defects in mRNA Translation in LRRK2-Mutant hiPSC-Derived Dopaminergic Neurons Lead to Dysregulated Calcium Homeostasis.
Cell stem cell (2020-08-28)
Jungwoo Wren Kim, Xiling Yin, Aanishaa Jhaldiyal, Mohammed Repon Khan, Ian Martin, Zhong Xie, Tamara Perez-Rosello, Manoj Kumar, Leire Abalde-Atristain, Jinchong Xu, Li Chen, Stephen M Eacker, D James Surmeier, Nicholas T Ingolia, Ted M Dawson, Valina L Dawson
ZUSAMMENFASSUNG
The G2019S mutation in leucine-rich repeat kinase 2 (LRRK2) is a common cause of familial Parkinson's disease (PD). This mutation results in dopaminergic neurodegeneration via dysregulated protein translation, although how alterations in protein synthesis contribute to neurodegeneration in human neurons is not known. Here we define the translational landscape in LRRK2-mutant dopaminergic neurons derived from human induced pluripotent stem cells (hiPSCs) via ribosome profiling. We found that mRNAs that have complex secondary structure in the 5' untranslated region (UTR) are translated more efficiently in G2019S LRRK2 neurons. This leads to the enhanced translation of multiple genes involved in Ca2+ regulation and to increased Ca2+ influx and elevated intracellular Ca2+ levels, a major contributor to PD pathogenesis. This study reveals a link between dysregulated translation control and Ca2+ homeostasis in G2019S LRRK2 human dopamine neurons, which potentially contributes to the progressive and selective dopaminergic neurotoxicity in PD.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Laminin aus Engelbreth-Holm-Swarm murine sarcoma basement membrane, 1-2 mg/mL in Tris-buffered saline, 0.2 μm filtered, BioReagent, suitable for cell culture
Roche
cOmplete, Mini, EDTA-freier Protease-Inhibitor-Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
Retinsäure, ≥98% (HPLC), powder
Sigma-Aldrich
N6,2′-O-Dibutyryladenosin 3′,5′-zyklisches Monophosphat Natriumsalz Monohydrat, ≥96% (HPLC), powder
Sigma-Aldrich
Actidion, from microbial, ≥94% (TLC)
Sigma-Aldrich
Y-27632 -dihydrochlorid, ≥98% (HPLC)
Sigma-Aldrich
Anti-Tyrosin-Hydroxylase-Antikörper, Chemicon®, from rabbit
Sigma-Aldrich
DAPT, ≥98% (HPLC), solid
Sigma-Aldrich
(+)-Natrium-L-ascorbat, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Purmorphamine, ≥98% (HPLC)
Sigma-Aldrich
Purmorphamin, A cell-permeable activator of Hedgehog signaling that induces osteoblast differentiation of multipotent mesenchymal progenitor cells C3H10T1/2 (EC50 = 1 µM).
Sigma-Aldrich
Isradipin, ≥98% (HPLC), solid