Merck
  • A novel amiloride-sensitive h+ transport pathway mediates enhanced superoxide production in thick ascending limb of salt-sensitive rats, not na+/h+ exchange.
A novel amiloride-sensitive h+ transport pathway mediates enhanced superoxide production in thick ascending limb of salt-sensitive rats, not na+/h+ exchange.
Hypertension (Dallas, Tex. : 1979) (2009-07-01)
Paul M O'Connor, Limin Lu, Mingyu Liang, Allen W Cowley
ZUSAMMENFASSUNG
It has been reported previously that H(+) efflux via the Na(+)/H(+) exchange stimulates NAD(P)H oxidase-dependent superoxide (O(2)(.-)) production in medullary thick ascending limb. We have demonstrated recently that N-methyl-amiloride-sensitive O(2)(.-) production is enhanced in the thick ascending limb of Dahl salt-sensitive (SS) rats, suggesting that H(+) efflux through Na(+)/H(+) exchangers may promote renal oxidative stress and the development of hypertension in these animals. In the current study we demonstrate, using selective and potent inhibitors, that inhibition of Na(+)/H(+) exchange does not mediate the ability of N-methyl-amiloride to inhibit thick ascending limb O(2)(.-) production. To determine the mechanism of action of N-methyl-amiloride, we examined H(+) efflux and O(2)(.-) production in SS and SS.13(BN) thick ascending limbs of prehypertensive, 0.4% NaCl-fed rats. Tissue strips containing the medullary thick ascending limb were isolated from male SS and salt-resistant consomic SS.13(BN) rats, loaded with either dihydroethedium or 2',7'-bis-(2-carboxyethyl)-5-(and-6)-carboxyfluorescein, acetoxymethyl ester, and imaged in a heated tissue bath. In Na(+)-replete media, activation of Na(+)/H(+) exchange using an NH(4)Cl prepulse did not stimulate thick ascending limb O(2)(.-) production. In Na(+)-free media containing BaCl(2) in which Na(+)/H(+) activity was inhibited, an NH(4)Cl prepulse stimulated O(2)(.-) production in medullary thick ascending limb renal tubular segments. This response was enhanced in medullary thick ascending limb of SS rats (slope Deltaethidium/Deltadihydroethedium=0.029+/-0.004) compared with SS.13(BN) rats (slope=0.010+/-0.004; P<0.04) and could be inhibited by N-methyl-amiloride (slope=0.005+/-0.002 and 0.006+/-0.002 for SS and SS.13(BN), respectively). We concluded that only H(+) efflux through a specific, as-yet-unidentified, amiloride-sensitive H(+) channel promotes O(2)(.-) production in the medullary thick ascending limb and that this channel is upregulated in SS rats.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)