Merck
  • Regulation of growth, intestinal microflora composition and expression of immune-related genes by dietary supplementation of Streptococcus faecalis in blunt snout bream (Megalobrama amblycephala).
Regulation of growth, intestinal microflora composition and expression of immune-related genes by dietary supplementation of Streptococcus faecalis in blunt snout bream (Megalobrama amblycephala).
Fish & shellfish immunology (2020-07-12)
Chao Xu, Xiao-Qun Zhong, Xiang-Fei Li, Hua-Juan Shi, Wen-Bin Liu
ZUSAMMENFASSUNG
A 10-week feeding trial was performed to investigate the effects of Streptococcus faecalis on the growth, intestinal microflora composition and expression of immune-related genes of blunt snout bream (Megalobrama amblycephala). Fish (46.32 ± 0.09 g) were fed four experimental diets containing 0 cfu/g (SF0, control), 1 × 105 cfu/g (SF1), 1 × 106 cfu/g (SF2) and 1 × 107 cfu/g (SF3) of S. faecalis, respectively. Results showed that daily growth index (DGI), feed efficiency ratio (FER), plasma glucose level, plasma contents of total protein and albumin as well as intestinal serous layer (SL), muscular layer (ML), submucous layer (SML), villi thickness (VT) and lamina propria (LP) were all no significant difference among all the treatments, whereas their (except plasma albumin content and intestinal ML) relatively high values were found in the SF2 group. Meanwhile, the intake of the SF2 diets significantly increased plasma globulin content and intestinal digestive enzymes activities, the opposite was true for the activities of plasma aspartate aminotransferase (AST) and alanine transaminase (ALT). In addition, the analysis of the intestinal microbiota showed that fish fed the SF2 diet have the highest values of intestinal alpha diversity and intestinal abundances of Actinobacteria, Chlamydiae, Firmicutes, Planctomycetes, Verrucomicrobia, Clostridium and Synechococcus, while the opposite was true for intestinal abundances of Acinetobacter, Anoxybacillus, Flavobacterium, Planctomyces, Plesiomonas, Pseudomonas, Staphylococcus and Clostridium perfringens. At the molecular level, the expression levels of tumour necrosis factor α (TNF α), interleukin 1β (IL 1β) and heat shock proteins 7 (HSP 70) in head kidney and spleen were all decreased significantly with the increasing S. faecalis levels up to 1 × 106 cfu/g, and then they were increased with further increasing S. faecalis levels. Overall, dietary supplementation of S. faecalis at 1 × 106 cfu/g could improve the intestinal health and innate immunity of blunt snout bream.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
S-Benzoylthiamine O-monophosphate