Merck
  • CD133/Src axis mediates tumor initiating property and epithelial-mesenchymal transition of head and neck cancer.
CD133/Src axis mediates tumor initiating property and epithelial-mesenchymal transition of head and neck cancer.
PloS one (2011-12-06)
Yu-Syuan Chen, Meng-Ju Wu, Chih-Yang Huang, Shu-Chun Lin, Tsung-Hsien Chuang, Cheng-Chia Yu, Jeng-Fan Lo
ZUSAMMENFASSUNG
Head and Neck squamous cell carcinoma (HNSCC) is a human lethal cancer with clinical, pathological, phenotypical and biological heterogeneity. Caner initiating cells (CICs), which are responsible for tumor growth and coupled with gain of epithelial-mesenchymal transition (EMT), have been identified. Previously, we enriched a subpopulation of head and neck cancer initiating cells (HN-CICs) with up-regulation of CD133 and enhancement of EMT. Others demonstrate that Src kinase interacts with and phosphorylates the cytoplasmic domain of CD133. However, the physiological function of CD133/Src signaling in HNSCCs has not been uncovered. Herein, we determined the critical role of CD133/Src axis modulating stemness, EMT and tumorigenicity of HNSCC and HN-CICs. Initially, down-regulation of CD133 significantly reduced the self-renewal ability and expression of stemness genes, and promoted the differentiation and apoptotic capability of HN-CICs. Additionally, knockdown of CD133 in HN-CICs also lessened both in vitro malignant properties including cell migration/cell invasiveness/anchorage independent growth, and in vivo tumor growth by nude mice xenotransplantation assay. In opposite, overexpression of CD133 enhanced the stemness properties and tumorigenic ability of HNSCCs. Lastly, up-regulation of CD133 increased phosphorylation of Src coupled with EMT transformation in HNSCCs, on the contrary, silence of CD133 or treatment of Src inhibitor inversely abrogated above phenotypic effects, which were induced by CD133 up-regulation in HNSCCs or HN-CICs. Our results suggested that CD133/Src signaling is a regulatory switch to gain of EMT and of stemness properties in HNSCC. Finally, CD133/Src axis might be a potential therapeutic target for HNSCC by eliminating HN-CICs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Agar, microbiology tested, suitable for plant cell culture, suitable for cell culture, powder
Sigma-Aldrich
Agar, suitable for plant cell culture, powder
Millipore
Agar, powder, suitable for microbiology
Sigma-Aldrich
Agar, Bacteriological, microbiology tested, suitable for plant cell culture, granular
Millipore
Agar, suitable for microbiology
Sigma-Aldrich
Agar, ash 2.0-4.5%
Sigma-Aldrich
Agar, High gel strength, suitable for plant cell culture
Sigma-Aldrich
Agar, Type A, suitable for plant cell culture
Sigma-Aldrich
Agar, Type M, suitable for plant cell culture
Millipore
Agar, suitable for microbiology
Sigma-Aldrich
Fumitremorgin C, from Neosartorya fischeri, film
Sigma-Aldrich
Agar, suitable for plant cell culture
Millipore
Agar, high purity, low ionic content, low gel strenght, suitable for microbiology