Merck
FAM46C/TENT5C functions as a tumor suppressor through inhibition of Plk4 activity.
Communications biology (2020-08-19)
Karineh Kazazian, Yosr Haffani, Deanna Ng, Chae Min Michelle Lee, Wendy Johnston, Minji Kim, Roland Xu, Karina Pacholzyk, Francis Si-Wah Zih, Julie Tan, Alannah Smrke, Aaron Pollett, Hannah Sun-Tsi Wu, Carol Jane Swallow
ZUSAMMENFASSUNG
Polo like kinase 4 (Plk4) is a tightly regulated serine threonine kinase that governs centriole duplication. Increased Plk4 expression, which is a feature of many common human cancers, causes centriole overduplication, mitotic irregularities, and chromosomal instability. Plk4 can also promote cancer invasion and metastasis through regulation of the actin cytoskeleton. Herein we demonstrate physical interaction of Plk4 with FAM46C/TENT5C, a conserved protein of unknown function until recently. FAM46C localizes to centrioles, inhibits Plk4 kinase activity, and suppresses Plk4-induced centriole duplication. Interference with Plk4 function by FAM46C was independent of the latter's nucleotidyl transferase activity. In addition, FAM46C restrained cancer cell invasion and suppressed MDA MB-435 cancer growth in a xenograft model, opposing the effect of Plk4. We demonstrate loss of FAM46C in patient-derived colorectal cancer tumor tissue that becomes more profound with advanced clinical stage. These results implicate FAM46C as a tumor suppressor that acts by inhibiting Plk4 activity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® shRNA, Bacterial Stock
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Triton X-100, laboratory grade
Millipore
ANTI-FLAG® M2-Affinitätsgel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
MISSION® shRNA, Purified DNA
Sigma-Aldrich
Anti-Centrin-Antikörper, Klon 20H5, clone 20H5, from mouse
Sigma-Aldrich
MISSION® Luciferase shRNA Control Vector, shRNA sequence targeting luciferase
Sigma-Aldrich
Anti-PLK-4 Antibody, clone 6H5, clone 6H5, from mouse
Sigma-Aldrich
MISSION® esiRNA, targeting human FAM46C