Merck
  • A comprehensive in vivo and mathematic modeling-based kinetic characterization for aspirin-induced chemoprevention in colorectal cancer.
A comprehensive in vivo and mathematic modeling-based kinetic characterization for aspirin-induced chemoprevention in colorectal cancer.
Carcinogenesis (2020-01-07)
Tadanobu Shimura, Shusuke Toden, Natalia L Komarova, Crichard Boland, Dominik Wodarz, Ajay Goel
ZUSAMMENFASSUNG
Accumulating evidence suggests that aspirin has anti-tumorigenic properties in colorectal cancer (CRC). Herein, we undertook a comprehensive and systematic series of in vivo animal experiments followed by 3D-mathematical modeling to determine the kinetics of aspirin's anti-cancer effects on CRC growth. In this study, CRC xenografts were generated using four CRC cell lines with and without PIK3CA mutations and microsatellite instability, and the animals were administered with various aspirin doses (0, 15, 50, and 100 mg/kg) for 2 weeks. Cell proliferation, apoptosis and protein expression were evaluated, followed by 3D-mathematical modeling analysis to estimate cellular division and death rates and their impact on aspirin-mediated changes on tumor growth. We observed that aspirin resulted in a dose-dependent decrease in the cell division rate, and a concomitant increase in the cell death rates in xenografts from all cell lines. Aspirin significantly inhibited cell proliferation as measured by Ki67 staining (P < 0.05-0.01). The negative effect of aspirin on the rate of tumor cell proliferation was more significant in xenograft tumors derived from PIK3CA mutant versus wild-type cells. A computational model of 3D-tumor growth suggests that the growth inhibitory effect of aspirin on the tumor growth kinetics is due to a reduction of tumor colony formation, and that this effect is sufficiently strong to be an important contributor to the reduction of CRC incidence in aspirin-treated patients. In conclusion, we provide a detailed kinetics of aspirin-mediated inhibition of tumor cell proliferation, which support the epidemiological data for the observed protective effect of aspirin in CRC patients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-15, ascites fluid
Sigma-Aldrich
Aspirin, meets USP testing specifications
Sigma-Aldrich
ApopTag Peroxidase In-situ-Apoptosenachweis-Kit, The ApopTag Peroxidase In Situ Apoptosis Detection Kit detects apoptotic cells in situ by labeling & detecting DNA strand breaks by the TUNEL method.