Merck
  • Predicting human serum albumin affinity of interleukin-8 (CXCL8) inhibitors by 3D-QSPR approach.
Predicting human serum albumin affinity of interleukin-8 (CXCL8) inhibitors by 3D-QSPR approach.
Journal of medicinal chemistry (2005-04-02)
Loretta Aureli, Gabriele Cruciani, Maria Candida Cesta, Roberto Anacardio, Lucio De Simone, Alessio Moriconi
ZUSAMMENFASSUNG
A novel class of 2-(R)-phenylpropionamides has been recently reported to inhibit in vitro and in vivo interleukin-8 (CXCL8)-induced biological activities. These CXCL8 inhibitors are derivatives of phenylpropionic nonsteroidal antiinflammatory drugs (NSAIDs), high-affinity ligands for site II of human serum albumin (HSA). Up to date, only a limited number of in silico models for the prediction of albumin protein binding are available. A three-dimensional quantitative structure-property relationship (3D-QSPR) approach was used to model the experimental affinity constant (K(i)) to plasma proteins of 37 structurally related molecules, using physicochemical and 3D-pharmacophoric descriptors. Molecular docking studies highlighted that training set molecules preferentially bind site II of HSA. The obtained model shows satisfactory statistical parameters both in fitting and predicting validation. External validation confirmed the statistical significance of the chemometric model, which is a powerful tool for the prediction of HSA binding in virtual libraries of structurally related compounds.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salicylsäure, ACS reagent, ≥99.0%
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications
Sigma-Aldrich
Furosemid
Sigma-Aldrich
Biphenyl-4-Carboxylsäure, 95%
Sigma-Aldrich
Salicylsäure, BioXtra, ≥99.0%
Sigma-Aldrich
Ketoprofen, ≥98% (TLC)
Sigma-Aldrich
Salicylsäure, ≥99%, FG
Sigma-Aldrich
Salicylsäure, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
4-Phenylphenol, 97%
Supelco
Ibuprofen
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Salicylsäure, suitable for plant cell culture
Supelco
Tolbutamid, analytical standard
Sigma-Aldrich
Salicylsäure, ReagentPlus®, ≥99%
Sigma-Aldrich
Salicylsäure, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
5-Chlorosalicylsäure, 98%
Sigma-Aldrich
Hydrochlorthiazid, crystalline
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Sulindac, ≥98.0%
Supelco
Salicylsäure, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Flurbiprofen, cyclooxygenase inhibitor
Sigma-Aldrich
Hydrochlorthiazid, meets USP testing specifications
Sigma-Aldrich
Ketoprofen, meets USP testing specifications
Sigma-Aldrich
4-Phenylphenol, purified by sublimation, 99%