Merck
TRIM24 facilitates antiviral immunity through mediating K63-linked TRAF3 ubiquitination.
The Journal of experimental medicine (2020-04-24)
Qingchen Zhu, Tao Yu, Shucheng Gan, Yan Wang, Yifei Pei, Qifan Zhao, Siyu Pei, Shumeng Hao, Jia Yuan, Jing Xu, Fajian Hou, Xuefeng Wu, Chao Peng, Ping Wu, Jun Qin, Yichuan Xiao
ZUSAMMENFASSUNG
Ubiquitination is an essential mechanism in the control of antiviral immunity upon virus infection. Here, we identify a series of ubiquitination-modulating enzymes that are modulated by vesicular stomatitis virus (VSV). Notably, TRIM24 is down-regulated through direct transcriptional suppression induced by VSV-activated IRF3. Reducing or ablating TRIM24 compromises type I IFN (IFN-I) induction upon RNA virus infection and thus renders mice more sensitive to VSV infection. Mechanistically, VSV infection induces abundant TRIM24 translocation to mitochondria, where TRIM24 binds with TRAF3 and directly mediates K63-linked TRAF3 ubiquitination at K429/K436. This modification of TRAF3 enables its association with MAVS and TBK1, which consequently activates downstream antiviral signaling. Together, these findings establish TRIM24 as a critical positive regulator in controlling the activation of antiviral signaling and describe a previously unknown mechanism of TRIM24 function.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonale ANTI-FLAG® M2-Peroxidase (HRP) in Maus hergestellte Antikörper, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O127:B8, purified by phenol extraction
Sigma-Aldrich
Anti-VSV Glycoprotein antibody, Mouse monoclonal, clone P5D4, purified from hybridoma cell culture