Merck
An RNA Repair Operon Regulated by Damaged tRNAs.
Cell reports (2020-12-29)
Kevin J Hughes, Xinguo Chen, A Maxwell Burroughs, L Aravind, Sandra L Wolin
ZUSAMMENFASSUNG
Many bacteria contain an RNA repair operon, encoding the RtcB RNA ligase and the RtcA RNA cyclase, that is regulated by the RtcR transcriptional activator. Although RtcR contains a divergent version of the CARF (CRISPR-associated Rossman fold) oligonucleotide-binding regulatory domain, both the specific signal that regulates operon expression and the substrates of the encoded enzymes are unknown. We report that tRNA fragments activate operon expression. Using a genetic screen in Salmonella enterica serovar Typhimurium, we find that the operon is expressed in the presence of mutations that cause tRNA fragments to accumulate. RtcA, which converts RNA phosphate ends to 2', 3'-cyclic phosphate, is also required. Operon expression and tRNA fragment accumulation also occur upon DNA damage. The CARF domain binds 5' tRNA fragments ending in cyclic phosphate, and RtcR oligomerizes upon binding these ligands, a prerequisite for operon activation. Our studies reveal a signaling pathway involving broken tRNAs and implicate the operon in tRNA repair.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)
Sigma-Aldrich
Mitomycin C aus Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Carbenicillin Dinatriumsalz, 89.0-100.5% anhydrous basis
Sigma-Aldrich
Methylmethansulfonat, 99%
Sigma-Aldrich
Bleomycinsulfat aus Streptomyces verticillus, 1.5-2.0 units/mg solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Tetracyclin -hydrochlorid, ≥95% (European Pharmacopoeia HPLC assay), meets EP testing specifications
Sigma-Aldrich
2,6-Diamino-pimelinsäure, ≥98% (TLC)
Sigma-Aldrich
2,6-Diamino-pimelinsäure, ≥95.0% (NT)