Merck
Mucus production stimulated by IFN-AhR signaling triggers hypoxia of COVID-19.
Cell research (2020-11-08)
Yuying Liu, Jiadi Lv, Jiangning Liu, Man Li, Jing Xie, Qi Lv, Wei Deng, Nannan Zhou, Yabo Zhou, Jiangping Song, Peng Wang, Chuan Qin, Wei-Min Tong, Bo Huang
ZUSAMMENFASSUNG
Silent hypoxia has emerged as a unique feature of coronavirus disease 2019 (COVID-19). In this study, we show that mucins are accumulated in the bronchoalveolar lavage fluid (BALF) of COVID-19 patients and are upregulated in the lungs of severe respiratory syndrome coronavirus 2 (SARS-CoV-2)-infected mice and macaques. We find that induction of either interferon (IFN)-β or IFN-γ upon SARS-CoV-2 infection results in activation of aryl hydrocarbon receptor (AhR) signaling through an IDO-Kyn-dependent pathway, leading to transcriptional upregulation of the expression of mucins, both the secreted and membrane-bound, in alveolar epithelial cells. Consequently, accumulated alveolar mucus affects the blood-gas barrier, thus inducing hypoxia and diminishing lung capacity, which can be reversed by blocking AhR activity. These findings potentially explain the silent hypoxia formation in COVID-19 patients, and suggest a possible intervention strategy by targeting the AhR pathway.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DL-Kynurenin, ≥95.0% (NT)
Sigma-Aldrich
Anti-IDO(Indolamin-2,3-Dioxygenase)-Antikörper, Klon 10.1, clone 10.1, Upstate®, from mouse
Sigma-Aldrich
Suplatast tosylate, ≥98% (HPLC)