Merck
Human PEPT1 pharmacophore distinguishes between dipeptide transport and binding.
Journal of medicinal chemistry (2006-06-09)
Balvinder S Vig, Terry R Stouch, Julita K Timoszyk, Yong Quan, Doris A Wall, Ronald L Smith, Teresa N Faria
ZUSAMMENFASSUNG
The human intestinal oligopeptide transporter (PEPT1) facilitates the absorption of dipeptides, tripeptides, and many peptidomimetic drugs. In this study, a large number of peptides were selected to investigate the structural features required for PEPT1 transport. Binding affinity was determined in a Gly-Sar uptake inhibition assay, whereas functional transport was ranked in a membrane depolarization assay. Although most of the peptides tested could bind to PEPT1, not all were substrates. As expected, single amino acids and tetrapeptides could not bind to or be transported by PEPT1. Dipeptide transport was influenced by charge, hydrophobicity, size, and side chain flexibility. The extent of transport was variable, and unexpectedly, some dipeptides were not substrates of PEPT1. These included dipeptides with two positive charges or extreme bulk in either position 1 or 2. Our results identify key features required for PEPT1 transport in contrast to most previously described pharmacophores, which are based on the inhibition of transport of a known substrate.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycin, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycin, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
L-Valin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
SAFC
Glycin
Sigma-Aldrich
Gly-Gly, ≥99% (titration)
Sigma-Aldrich
L-Valin, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Glycin, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycin, ACS reagent, ≥98.5%
Sigma-Aldrich
Gly-Gly-Gly
Sigma-Aldrich
L-Valin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Glycin, 99%, FCC
Sigma-Aldrich
Glycin, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycin, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycin 1 M -Lösung
Sigma-Aldrich
Ala-Ala
Sigma-Aldrich
Gly-Tyr
Sigma-Aldrich
Gly-Gly, BioPerformance Certified, suitable for cell culture, ≥99%
Sigma-Aldrich
Gly-Gly, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Gly-Sar
Supelco
Glycin, analytical standard, for nitrogen determination according to Kjeldahl method
Sigma-Aldrich
Gly-Phe
Sigma-Aldrich
Gly-Gly-Gly, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Gly-Glu
Sigma-Aldrich
Gly-Gly-Gly-Gly
Sigma-Aldrich
Glycin, tested according to Ph. Eur.
Supelco
L-Valin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland