Merck
Investigating ultrasound-light interaction in scattering media.
Journal of biomedical optics (2020-02-28)
Yujia Huang, Michelle Cua, Joshua Brake, Yan Liu, Changhuei Yang
ZUSAMMENFASSUNG
Ultrasound-assisted optical imaging techniques, such as ultrasound-modulated optical tomography, allow for imaging deep inside scattering media. In these modalities, a fraction of the photons passing through the ultrasound beam is modulated. The efficiency by which the photons are converted is typically referred to as the ultrasound modulation's "tagging efficiency." Interestingly, this efficiency has been defined in varied and discrepant fashion throughout the scientific literature. The aim of this study is the ultrasound tagging efficiency in a manner consistent with its definition and experimentally verify the contributive (or noncontributive) relationship between the mechanisms involved in the ultrasound optical modulation process. We adopt a general description of the tagging efficiency as the fraction of photons traversing an ultrasound beam that is frequency shifted (inclusion of all frequency-shifted components). We then systematically studied the impact of ultrasound pressure and frequency on the tagging efficiency through a balanced detection measurement system that measured the power of each order of the ultrasound tagged light, as well as the power of the unmodulated light component. Through our experiments, we showed that the tagging efficiency can reach 70% in a scattering phantom with a scattering anisotropy of 0.9 and a scattering coefficient of 4  mm  -  1 for a 1-MHz ultrasound with a relatively low (and biomedically acceptable) peak pressure of 0.47 MPa. Furthermore, we experimentally confirmed that the two ultrasound-induced light modulation mechanisms, particle displacement and refractive index change, act in opposition to each other. Tagging efficiency was quantified via simulation and experiments. These findings reveal avenues of investigation that may help improve ultrasound-assisted optical imaging techniques.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Gelatine aus Schweinehaut, gel strength 300, Type A
Sigma-Aldrich
Phenyl-bis(2,4,6-trimethylbenzoyl)phosphinoxid, 97%, powder
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Hämin, from bovine, ≥90%
Sigma-Aldrich
Hämin, BioXtra, from Porcine, ≥96.0% (HPLC)
Sigma-Aldrich
Flavin-Adenin-Dinukleotid Dinatriumsalz Hydrat, ≥95% (HPLC), powder
Sigma-Aldrich
Carrageenan, suitable for gel preparation
Sigma-Aldrich
Protoporphyrin-IX, ≥95%
Supelco
Phosphorpentoxid, ≥99.99% trace metals basis
Sigma-Aldrich
Pentostatin, ≥95% (HPLC)