Merck
  • Structure-activity relationships of alpha-amino acid ligands for the alpha2delta subunit of voltage-gated calcium channels.
Structure-activity relationships of alpha-amino acid ligands for the alpha2delta subunit of voltage-gated calcium channels.
Bioorganic & medicinal chemistry letters (2005-12-29)
Kathleen H Mortell, David J Anderson, James J Lynch, Sherry L Nelson, Kathy Sarris, Heath McDonald, Reza Sabet, Scott Baker, Prisca Honore, Chih-Hung Lee, Michael F Jarvis, Murali Gopalakrishnan
ZUSAMMENFASSUNG
A series of alpha-amino acids were identified as ligands which compete with gabapentin for binding to the alpha(2)delta subunit of voltage-dependent Ca(2+) channels. Potent analogs were identified. Their activity in an in vivo pain assay is described.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Phenylalanin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Phenylalanin, reagent grade, ≥98%
Sigma-Aldrich
L-Phenylalanin, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
D−(−)-α-Phenylglycin, 99%
Sigma-Aldrich
Gabapentin, solid
Sigma-Aldrich
L−(+)-α-Phenylglycin, 99%
Sigma-Aldrich
L-Phenylalanin, 99%, FCC
Supelco
L-Phenylalanin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Ethionin, ≥99% (TLC)
Sigma-Aldrich
DL-Homophenylalanin, 98%