Merck
CTCF-promoted RNA polymerase II pausing links DNA methylation to splicing.
Nature (2011-10-04)
Sanjeev Shukla, Ersen Kavak, Melissa Gregory, Masahiko Imashimizu, Bojan Shutinoski, Mikhail Kashlev, Philipp Oberdoerffer, Rickard Sandberg, Shalini Oberdoerffer
ZUSAMMENFASSUNG
Alternative splicing of pre-messenger RNA is a key feature of transcriptome expansion in eukaryotic cells, yet its regulation is poorly understood. Spliceosome assembly occurs co-transcriptionally, raising the possibility that DNA structure may directly influence alternative splicing. Supporting such an association, recent reports have identified distinct histone methylation patterns, elevated nucleosome occupancy and enriched DNA methylation at exons relative to introns. Moreover, the rate of transcription elongation has been linked to alternative splicing. Here we provide the first evidence that a DNA-binding protein, CCCTC-binding factor (CTCF), can promote inclusion of weak upstream exons by mediating local RNA polymerase II pausing both in a mammalian model system for alternative splicing, CD45, and genome-wide. We further show that CTCF binding to CD45 exon 5 is inhibited by DNA methylation, leading to reciprocal effects on exon 5 inclusion. These findings provide a mechanistic basis for developmental regulation of splicing outcome through heritable epigenetic marks.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Normales Maus-IgG, Normal Mouse IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.
Sigma-Aldrich
Anti-RNA-Polymerase II-Antikörper, Klon CTD4H8, clone CTD4H8, Upstate®, from mouse
Sigma-Aldrich
Anti-CTCF-Antikörper, serum, Upstate®
Sigma-Aldrich
Anti-Trimethyl-Histon-H3(-Lys4)-Antikörper, Klon MC315, monoklonaler Kaninchen-Antikörper, culture supernatant, clone MC315, Upstate®