Merck
  • Identification of a short linear sequence present in the C-terminal tail of the rat follitropin receptor that modulates arrestin-3 binding in a phosphorylation-independent fashion.
Identification of a short linear sequence present in the C-terminal tail of the rat follitropin receptor that modulates arrestin-3 binding in a phosphorylation-independent fashion.
The Journal of biological chemistry (2002-04-06)
Hiroshi Kishi, Hanumanthappa Krishnamurthy, Colette Galet, Ravi Sankar Bhaskaran, Mario Ascoli
ZUSAMMENFASSUNG
The rat follitropin receptor (rFSHR) is an unusual G protein-coupled receptor in that agonist-induced activation leads to the phosphorylation of the first and third intracellular loops instead of the C-terminal tail. To determine regions of G protein-coupled receptors that affect internalization independently of phosphorylation we examined the effects of truncations of the C-terminal tail of the rFSHR on agonist-induced internalization. Our studies show that progressive truncations of a region flanked by residues 642 and 651 enhance the internalization of human follicle-stimulating hormone (hFSH). Further characterization of a mutant truncated at residue 649 (designated rFSHR-t649) and another mutant in which the 642-651 region was deleted in the context of the full-length rFSHR, designated rFSHR(Delta642-651), showed that both of them internalized hFSH at rates that were 2-3 times faster than rFSHR-wild type (wt). Like rFSHR-wt, however, the internalization of hFSH mediated by rFSHR-t649 and rFSHR(Delta642-651) can be inhibited with dominant-negative mutants of the non-visual arrestins or dynamin. Alanine-scanning mutagenesis of the 642-651 region suggests that the effects on internalization are not mediated by a single residue, however. In an attempt to understand the molecular basis of the enhanced internalization of hFSH mediated by these mutants we used an assay that can be readily used to assess the association of the rFSHR with the arrestin-3 in co-transfected cells. Using this assay we were able to show that, when compared with rFSHR-wt, rFSHR(Delta642-651) displays an approximately 4-fold enhancement in binding affinity for arrestin-3 and an approximately 1.7-fold reduction in maximal arrestin-3 binding capacity. We conclude that a short linear sequence present in the C-terminal tail of the rFSHR (642SATHNFHARK651) that is not phosphorylated limits internalization by lowering the affinity of the rFSHR for the endogenous non-visual arrestins.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonale ANTI-FLAG® M2-Peroxidase (HRP) in Maus hergestellte Antikörper, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Myc-Tag-Antikörper, Klon 9E10, clone 9E10, Upstate®, from mouse