Merck
  • Prevention of dsRNA-induced interferon signaling by AGO1x is linked to breast cancer cell proliferation.
Prevention of dsRNA-induced interferon signaling by AGO1x is linked to breast cancer cell proliferation.
The EMBO journal (2020-08-20)
Souvik Ghosh, Joao C Guimaraes, Manuela Lanzafame, Alexander Schmidt, Afzal Pasha Syed, Beatrice Dimitriades, Anastasiya Börsch, Shreemoyee Ghosh, Nitish Mittal, Thomas Montavon, Ana Luisa Correia, Johannes Danner, Gunter Meister, Luigi M Terracciano, Sébastien Pfeffer, Salvatore Piscuoglio, Mihaela Zavolan
ZUSAMMENFASSUNG
Translational readthrough, i.e., elongation of polypeptide chains beyond the stop codon, was initially reported for viral RNA, but later found also on eukaryotic transcripts, resulting in proteome diversification and protein-level modulation. Here, we report that AGO1x, an evolutionarily conserved translational readthrough isoform of Argonaute 1, is generated in highly proliferative breast cancer cells, where it curbs accumulation of double-stranded RNAs (dsRNAs) and consequent induction of interferon responses and apoptosis. In contrast to other mammalian Argonaute protein family members with primarily cytoplasmic functions, AGO1x exhibits nuclear localization in the vicinity of nucleoli. We identify AGO1x interaction with the polyribonucleotide nucleotidyltransferase 1 (PNPT1) and show that the depletion of this protein further augments dsRNA accumulation. Our study thus uncovers a novel function of an Argonaute protein in buffering the endogenous dsRNA-induced interferon responses, different than the canonical function of AGO proteins in the miRNA effector pathway. As AGO1x expression is tightly linked to breast cancer cell proliferation, our study thus suggests a new direction for limiting tumor growth.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Triton X-100, Molecular Biology
Millipore
Anti-FLAG® M2 Magnetperlen, affinity isolated antibody
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Esel-Serum
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
TRI-Reagenz®
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)
Sigma-Aldrich
Anti-γ-Tubulin-Antikörper, monoklonaler Antikörper der Maus in Maus hergestellte Antikörper, clone GTU-88, ascites fluid
Sigma-Aldrich
Monoklonale Anti-GAPDH−-Peroxidase in Maus hergestellte Antikörper, clone GAPDH-71.1, purified immunoglobulin, buffered aqueous solution