Merck
  • Sickle cell disease mice have cerebral oxidative stress and vascular and white matter abnormalities.
Sickle cell disease mice have cerebral oxidative stress and vascular and white matter abnormalities.
Blood cells, molecules & diseases (2020-09-15)
Alfia Khaibullina, Luis E F Almeida, Sayuri Kamimura, Patricia M Zerfas, Meghann L Smith, Sebastian Vogel, Paul Wakim, Olavo M Vasconcelos, Martha M Quezado, Iren Horkayne-Szakaly, Zenaide M N Quezado, Alfia Khaibullina, Luis E F Almeida, Sayuri Kamimura, Patricia M Zerfas, Meghann L Smith, Sebastian Vogel, Paul Wakim, Olavo M Vasconcelos, Martha M Quezado, Iren Horkayne-Szakaly, Zenaide M N Quezado
ZUSAMMENFASSUNG
Strokes are feared complications of sickle cell disease (SCD) and yield significant neurologic and neurocognitive deficits. However, even without detectable strokes, SCD patients have significant neurocognitive deficits in domains of learning and memory, processing speed and executive function. In these cases, mechanisms unrelated to major cerebrovascular abnormalities likely underlie these deficits. While oxidative stress and stress-related signaling pathways play a role in SCD pathophysiology, their role in cerebral injury remains unknown. We have shown that Townes and BERK SCD mice, while not having strokes, recapitulate neurocognitive deficits reported in humans. We hypothesized that cognitive deficits in SCD mice are associated with cerebral oxidative stress. We showed that SCD mice have increased levels of reactive oxygen species, protein carbonylation, and lipid peroxidation in hippocampus and cortex, thus suggesting increased cerebral oxidative stress. Further, cerebral oxidative stress was associated with caspase-3 activity alterations and vascular endothelial abnormalities, white matter changes, and disruption of the blood brain barrier, similar to those reported after ischemic/oxidative injury. Additionally, after repeated hypoxia/reoxygenation exposure, homozygous Townes had enhanced microglia activation. Our findings indicate that oxidative stress and stress-induced tissue damage is increased in susceptible brain regions, which may, in turn, contribute to neurocognitive deficits in SCD mice.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Caspase-3-Assay-Kit, fluorimetrisch