Merck
  • Hypothalamic Pomc Neurons Innervate the Spinal Cord and Modulate the Excitability of Premotor Circuits.
Hypothalamic Pomc Neurons Innervate the Spinal Cord and Modulate the Excitability of Premotor Circuits.
Current biology : CB (2020-09-26)
Philip Reinoß, Elisa Ciglieri, Marielle Minére, Stephan Bremser, Andreas Klein, Heiko Löhr, Patrick M Fuller, Ansgar Büschges, Peter Kloppenburg, Henning Fenselau, Matthias Hammerschmidt
ZUSAMMENFASSUNG
Locomotion requires energy, yet animals need to increase locomotion in order to find and consume food in energy-deprived states. While such energy homeostatic coordination suggests brain origin, whether the central melanocortin 4 receptor (Mc4r) system directly modulates locomotion through motor circuits is unknown. Here, we report that hypothalamic Pomc neurons in zebrafish and mice have long-range projections into spinal cord regions harboring Mc4r-expressing V2a interneurons, crucial components of the premotor networks. Furthermore, in zebrafish, Mc4r activation decreases the excitability of spinal V2a neurons as well as swimming and foraging, while systemic or V2a neuron-specific blockage of Mc4r promotes locomotion. In contrast, in mice, electrophysiological recordings revealed that two-thirds of V2a neurons in lamina X are excited by the Mc4r agonist α-MSH, and acute inhibition of Mc4r signaling reduces locomotor activity. In addition, we found other Mc4r neurons in spinal lamina X that are inhibited by α-MSH, which is in line with previous studies in rodents where Mc4r agonists reduced locomotor activity. Collectively, our studies identify spinal V2a interneurons as evolutionary conserved second-order neurons of the central Mc4r system, providing a direct anatomical and functional link between energy homeostasis and locomotor control systems. The net effects of this modulatory system on locomotor activity can vary between different vertebrate species and, possibly, even within one species. We discuss the biological sense of this phenomenon in light of the ambiguity of locomotion on energy balance and the different living conditions of the different species.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
α-Melanozyten-stimulierendes Hormon, ≥97% (HPLC)
Sigma-Aldrich
Dextransulfat Natriumsalz aus Leuconostoc spp., Molecular Biology, average Mw >500,000 (dextran starting material), contains 0.5-2% phosphate buffer
Sigma-Aldrich
Biocytin, ≥98% (TLC)
Sigma-Aldrich
Amphotericin B aus Streptomyces sp., ~80% (HPLC), powder
Sigma-Aldrich
N-Methyl-D-Asparaginsäure, ≥98% (TLC), solid
Sigma-Aldrich
Ribonukleinsäure aus Torulahefe, Type VI
Sigma-Aldrich
Picrotoxin, powder
Sigma-Aldrich
Melanotan II acetate salt, ≥95% (HPLC)
Sigma-Aldrich
CNQX, ≥98% (HPLC), solid
Sigma-Aldrich
Strychnine hydrochloride
Supelco
Tolbutamid, analytical standard
Sigma-Aldrich
Tubocurarin, ≥97%