Merck
  • 3D in situ imaging of the female reproductive tract reveals molecular signatures of fertilizing spermatozoa in mice.
3D in situ imaging of the female reproductive tract reveals molecular signatures of fertilizing spermatozoa in mice.
eLife (2020-10-21)
Lukas Ded, Jae Yeon Hwang, Kiyoshi Miki, Huanan F Shi, Jean-Ju Chung
ZUSAMMENFASSUNG
Out of millions of ejaculated sperm, a few reach the fertilization site in mammals. Flagellar Ca2+ signaling nanodomains, organized by multi-subunit CatSper calcium channel complexes, are pivotal for sperm migration in the female tract, implicating CatSper-dependent mechanisms in sperm selection. Here using biochemical and pharmacological studies, we demonstrate that CatSper1 is an O-linked glycosylated protein, undergoing capacitation-induced processing dependent on Ca2+ and phosphorylation cascades. CatSper1 processing correlates with protein tyrosine phosphorylation (pY) development in sperm cells capacitated in vitro and in vivo. Using 3D in situ molecular imaging and ANN-based automatic detection of sperm distributed along the cleared female tract, we demonstrate that spermatozoa past the utero-tubal junction possess the intact CatSper1 signals. Together, we reveal that fertilizing mouse spermatozoa in situ are characterized by intact CatSper channel, lack of pY, and reacted acrosomes. These findings provide molecular insight into sperm selection for successful fertilization in the female reproductive tract.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kalzium-Ionophor A23187, ≥98% (TLC), powder
Sigma-Aldrich
N-Methyl-D-Glucamin, 99.0-100.5% (titration)
Millipore
Monoklonale Anti-HA−Agarose in Maus hergestellte Antikörper, clone HA-7, purified immunoglobulin, PBS suspension
Sigma-Aldrich
Anti-Phosphotyrosin-Antikörper, Klon 4G10®, clone 4G10®, Upstate®, from mouse
Sigma-Aldrich
Calyculin A, Discodermia calyx, Calyculin A, CAS 101932-71-2, is a cell-permeable inhibitor of protein phosphatase 2A (IC50 = 0.5-1 nM) and protein phosphatase 1 (PP1; IC50 = 2 nM).
Sigma-Aldrich
Hexafluorpropen, Trimer
Sigma-Aldrich
ALLN, Cell-permeable inhibitor of calpain I (Ki = 190 nM), calpain II (Ki = 220 nM), cathepsin B (Ki = 150 nM), and cathepsin L (Ki = 500 pM).