- Discovery and synthesis of new immunosuppressive alkaloids from the stem of Fissistigma oldhamii (Hemsl.) Merr.
Discovery and synthesis of new immunosuppressive alkaloids from the stem of Fissistigma oldhamii (Hemsl.) Merr.
Bioorganic & medicinal chemistry (2006-11-04)
Yi-Nan Zhang, Xiang-Gen Zhong, Zong-Ping Zheng, Xu-Dong Hu, Jian-Ping Zuo, Li-Hong Hu
PMID17081761
ZUSAMMENFASSUNG
Three new alkaloids (1-3) and twenty-one known compounds were isolated from the stem of Fissistigma oldhamii (Hemsl.) Merr. which was the ruler herb in an approved Traditional Chinese herbal formula used for treatment of rheumatoid arthritis in China and synthesis of one new immunosuppressive alkaloid was achieved. These compounds, including the crude extracts of this herb, exhibited strong activities in the inhibition of T and B cell proliferation.