Merck
  • Apoptotic regulation by the Crk adapter protein mediated by interactions with Wee1 and Crm1/exportin.
Apoptotic regulation by the Crk adapter protein mediated by interactions with Wee1 and Crm1/exportin.
Molecular and cellular biology (2002-02-13)
Jesse J Smith, D Ashley Richardson, Jan Kopf, Minoru Yoshida, Robert E Hollingsworth, Sally Kornbluth
ZUSAMMENFASSUNG
The adapter protein Crk contains an SH2 domain and two SH3 domains. Through binding of particular ligands to the SH2 domain and the N-terminal SH3 domain, Crk has been implicated in a number of signaling processes, including regulation of cell growth, cell motility, and apoptosis. We report here that the C-terminal SH3 domain, never shown to bind any specific signaling molecules, contains a binding site for the nuclear export factor Crm1. We find that a mutant Crk protein, deficient in Crm1 binding, promotes apoptosis. Moreover, this nuclear export sequence mutant [NES(-) Crk] interacts strongly, through its SH2 domain, with the nuclear tyrosine kinase, Wee1. Collectively, these data suggest that a nuclear population of Crk bound to Wee1 promotes apoptotic death of mammalian cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Phosphotyrosin-Antikörper, Klon 4G10®, clone 4G10®, Upstate®, from mouse