Merck
  • Structural requirements for drug inhibition of the liver specific human organic cation transport protein 1.
Structural requirements for drug inhibition of the liver specific human organic cation transport protein 1.
Journal of medicinal chemistry (2008-09-16)
Gustav Ahlin, Johan Karlsson, Jenny M Pedersen, Lena Gustavsson, Rolf Larsson, Pär Matsson, Ulf Norinder, Christel A S Bergström, Per Artursson
ZUSAMMENFASSUNG
The liver-specific organic cation transport protein (OCT1; SLC22A1) transports several cationic drugs including the antidiabetic drug metformin and the anticancer agents oxaliplatin and imatinib. In this study, we explored the chemical space of registered oral drugs with the aim of studying the inhibition pattern of OCT1 and of developing predictive computational models of OCT1 inhibition. In total, 191 structurally diverse compounds were examined in HEK293-OCT1 cells. The assay identified 47 novel inhibitors and confirmed 15 previously known inhibitors. The enrichment of OCT1 inhibitors was seen in several drug classes including antidepressants. High lipophilicity and a positive net charge were found to be the key physicochemical properties for OCT1 inhibition, whereas a high molecular dipole moment and many hydrogen bonds were negatively correlated to OCT1 inhibition. The data were used to generate OPLS-DA models for OCT1 inhibitors; the final model correctly predicted 82% of the inhibitors and 88% of the noninhibitors of the test set.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
N-Acetyl-L-Cystein, BioReagent, suitable for cell culture
Sigma-Aldrich
N-Acetyl-L-Cystein, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
Koffein, powder, ReagentPlus®
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Colchicin, ≥95% (HPLC), powder
Sigma-Aldrich
3,4-Dihydroxy-L-Phenylalanin, ≥98% (TLC)
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Folsäure, ≥97%
Sigma-Aldrich
β-Estradiol, ≥98%
Sigma-Aldrich
(−)-Nikotin, ≥99% (GC), liquid
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Theophyllin, anhydrous, ≥99%, powder
Sigma-Aldrich
Acetylsalicylsäure, ≥99.0%
Sigma-Aldrich
(−)-Epinephrin
Sigma-Aldrich
Trimethoprim, ≥98.5%
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Folsäure, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
Progesteron, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Koffein, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Tetracyclin, 98.0-102.0% (HPLC)
Sigma-Aldrich
Cysteamin, ~95%
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
Corticosteron, ≥98.5% (HPLC)
Sigma-Aldrich
Colchicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC)
Sigma-Aldrich
Histamin, ≥97.0%