Merck
  • Clozapine, nimodipine and endosulfan differentially suppress behavioral defects caused by gain-of-function mutations in a two-pore domain K+ channel (UNC-58).
Clozapine, nimodipine and endosulfan differentially suppress behavioral defects caused by gain-of-function mutations in a two-pore domain K+ channel (UNC-58).
Neuroscience research (2020-07-19)
Merve Kasap, Donard S Dwyer
ZUSAMMENFASSUNG
Two-pore domain K+ channels (K2Ps) regulate the resting membrane potential in excitable cells and determine ease of depolarization. Gain-of-function (gf) mutations in one of these channels (unc-58) in C. elegans switch it to a Na+ conductance channel and cause tremors, paralysis and other defects. We hypothesized that it should be possible to identify drugs that corrected these defects in unc-58(gf) mutant animals by blocking or modulating the over-active channels. We examined dispersal of animals on food because the absence of effective forward locomotion is the most obvious defect. In addition, we quantified egg release over 24 h. Starting with a known inhibitor of mammalian K2Ps and directed structure-based screening, we evaluated numerous drugs in these assays. Loratadine, which inhibits human KCNK18, significantly improved movement as did methiothepin. We confirmed that endosulfan, a GABA-A receptor antagonist, corrected locomotion in the unc-58(gf) strains. Based on structural similarities to other hits, we found that clozapine, loxapine and amoxapine potently suppressed abnormal phenotypes. Curiously, nimodipine, a Ca++-channel blocker, dramatically improved movement and egg laying in unc-58(e665), but not unc-58(n495) animals. Molecular modeling provided initial insights into a possible basis for this difference based on the location of the e665 and n495 mutations. This research may lead to identification of novel K2P modulators and potential leads for drug discovery.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Amoxapin