Merck
  • O-cyclic phytosphingosine-1-phosphate stimulates HIF1α-dependent glycolytic reprogramming to enhance the therapeutic potential of mesenchymal stem cells.
O-cyclic phytosphingosine-1-phosphate stimulates HIF1α-dependent glycolytic reprogramming to enhance the therapeutic potential of mesenchymal stem cells.
Cell death & disease (2019-08-07)
Hyun Jik Lee, Young Hyun Jung, Gee Euhn Choi, Jun Sung Kim, Chang Woo Chae, Jae Ryong Lim, Seo Yihl Kim, Joo Eun Lee, Min Chul Park, Jee Hyeon Yoon, Myeong Jun Choi, Kye-Seong Kim, Ho Jae Han
ZUSAMMENFASSUNG
O-cyclic phytosphingosine-1-phosphate (cP1P) is a novel chemically synthesized sphingosine metabolite derived from phytosphingosine-1-phosphate. Although structurally similar to sphingosine-1-phosphate (S1P), its biological properties in stem cells remain to be reported. We investigated the effect of cP1P on the therapeutic potential of mesenchymal stem cells (MSCs) and their regulatory mechanism. We found that, under hypoxia, cP1P suppressed MSC mitochondrial dysfunction and apoptosis. Metabolic data revealed that cP1P stimulated glycolysis via the upregulation of glycolysis-related genes. cP1P-induced hypoxia-inducible factor 1 alpha (HIF1α) plays a key role for MSC glycolytic reprogramming and transplantation efficacy. The intracellular calcium-dependent PKCα/mammalian target of the rapamycin (mTOR) signaling pathway triggered by cP1P regulated HIF1α translation via S6K1, which is critical for HIF1 activation. Furthermore, the cP1P-activated mTOR pathway induced bicaudal D homolog 1 expression, leading to HIF1α nuclear translocation. In conclusion, cP1P enhances the therapeutic potential of MSC through mTOR-dependent HIF1α translation and nuclear translocation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Duolink® In Situ Detektionsreagenzien Rot
Sigma-Aldrich
Cycloheximid -Lösung, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
Duolink® In Situ PLA® Sonde Anti-Maus PLUS
Sigma-Aldrich
Duolink® In Situ PLA® Sonde Anti-Kaninchen MINUS
Sigma-Aldrich
BAPTA-AM, ≥95% (HPLC)
Sigma-Aldrich
Rapamycin aus Streptomyces hygroscopicus, ≥95% (HPLC), powder
Sigma-Aldrich
Tetramethylrhodamin-Ethylester-Perchlorat, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Alkaline Phosphatase Diethanolamine Activity Kit, sufficient for 100 tests
Sigma-Aldrich
Akt-Inhibitor, The Akt Inhibitor controls the biological activity of Akt. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.