Merck
  • erm(B)-carrying elements in tetracycline-resistant pneumococci and correspondence between Tn1545 and Tn6003.
erm(B)-carrying elements in tetracycline-resistant pneumococci and correspondence between Tn1545 and Tn6003.
Antimicrobial agents and chemotherapy (2008-02-21)
Ileana Cochetti, Emily Tili, Marina Mingoia, Pietro E Varaldo, Maria Pia Montanari
ZUSAMMENFASSUNG
This study investigated the genetic organization of erm(B)-carrying transposons of Streptococcus pneumoniae and their distribution in tetracycline-resistant clinical isolates. By comparatively analyzing reference pneumococci carrying erm(B)/tet(M) transposon Tn1545, Tn6003, Tn6002, or Tn3872, we demonstrated a substantial correspondence between Tn1545 and Tn6003, which have the same resistance gene combination [tet(M) (tetracycline), erm(B) (erythromycin), and aphA-3 (kanamycin)]; share the macrolide-aminoglycoside-streptothricin element, containing a second erm(B); and only differ by a ca. 1.2-kb insertion (containing a putative IS1239 insertion sequence) detected in Tn1545 from S. pneumoniae reference strain BM4200. These results enabled elucidation of the structure of Tn1545, the first erm(B)-carrying transposon described in S. pneumoniae. A collection of 83 erythromycin- and tetracycline-resistant clinical pneumococci, representative of recent Italian isolates carrying erm(B) as the sole erythromycin resistance gene, was used to investigate the distribution of the different transposons. All 83 organisms were positive for tet(M) and bore an erm(B)/tet(M) transposon that could be characterized by using a specific set of primer pairs; Tn3872 was detected in 18 isolates, Tn6002 in 59 isolates, and Tn6003 in 6 (the sole kanamycin-resistant) isolates. The genetic organization of transposon Tn1545, with its specific insertion, was not detected in any of the isolates tested. The erm(B)-carrying elements of tetracycline-resistant pneumococci substantially corresponded to those [bearing a silent tet(M) gene] recently detected in tetracycline-susceptible pneumococci. Overall, in erm(B)-positive pneumococci, Tn6003 was the least common erm(B)-carrying Tn916-related element and Tn6002 the most common.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kanamycinsulfat, mixture of Kanamycin A (main component) and Kanamycin B and C
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Tetracyclin, 98.0-102.0% (HPLC)
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.