- Are all the DNA gyrase mutations found in Mycobacterium leprae clinical strains involved in resistance to fluoroquinolones?
Are all the DNA gyrase mutations found in Mycobacterium leprae clinical strains involved in resistance to fluoroquinolones?
Antimicrobial agents and chemotherapy (2007-12-12)
Stéphanie Matrat, Emmanuelle Cambau, Vincent Jarlier, Alexandra Aubry
PMID18070975
ZUSAMMENFASSUNG
Mycobacterium leprae DNA gyrases carrying various mutations, previously described in clinical strains, were investigated for quinolone susceptibility by inhibition of supercoiling and DNA cleavage promotion. We demonstrated that the gyrA mutations leading to G89C or A91V confer fluoroquinolone resistance whereas the gyrB mutation leading to D205N does not.