Merck
  • Carbonic anhydrase inhibitors. Cloning, characterization, and inhibition studies of a new beta-carbonic anhydrase from Mycobacterium tuberculosis.
Carbonic anhydrase inhibitors. Cloning, characterization, and inhibition studies of a new beta-carbonic anhydrase from Mycobacterium tuberculosis.
Journal of medicinal chemistry (2009-04-03)
Isao Nishimori, Tomoko Minakuchi, Daniela Vullo, Andrea Scozzafava, Alessio Innocenti, Claudiu T Supuran
ZUSAMMENFASSUNG
The Rv3273 gene product of Mycobacterium tuberculosis, a beta-carbonic anhydrase (CA, EC 4.2.1.1), mtCA 3, shows appreciable catalytic activity for CO(2) hydration (k(cat) of 4.3 x 10(5) s(-1), and k(cat)/K(m) of 4.0 x 10(7) M(-1) x s(-1)). A series of sulfonamides/sulfamates was assayed for their interaction with mtCA 3. Sulfanilyl-sulfonamides, acetazolamide, methazolamide, ethoxzolamide, dichlorophenamide, dorzolamide, brinzolamide, benzolamide, and zonisamide, showed effective, submicromolar inhibition (K(I)s of 104-611 nM), the best inhibitor being 2-amino-pyrimidin-4-yl-sulfanilamide (K(I) of 91 nM).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Sulfanilamid, ≥98%
Sigma-Aldrich
p-Toluolsulfonamid, ReagentPlus®, ≥99%
Sigma-Aldrich
Saccharin, ≥98%
Sigma-Aldrich
Saccharin, ≥99%
Sigma-Aldrich
Sulfanilamid, puriss. p.a., ≥98% (calc. to the dried substance)
Sigma-Aldrich
p-Toluolsulfonamid, reagent grade, 97%
Sigma-Aldrich
4-Sulfamoylbenzoesäure, 97%
Sigma-Aldrich
(±)-Sulpirid
Supelco
Sulfanilamid, VETRANAL®, analytical standard