Merck
Four Key Steps Control Glycolytic Flux in Mammalian Cells.
Cell systems (2018-07-02)
Lukas Bahati Tanner, Alexander G Goglia, Monica H Wei, Talen Sehgal, Lance R Parsons, Junyoung O Park, Eileen White, Jared E Toettcher, Joshua D Rabinowitz
ZUSAMMENFASSUNG
Altered glycolysis is a hallmark of diseases including diabetes and cancer. Despite intensive study of the contributions of individual glycolytic enzymes, systems-level analyses of flux control through glycolysis remain limited. Here, we overexpress in two mammalian cell lines the individual enzymes catalyzing each of the 12 steps linking extracellular glucose to excreted lactate, and find substantial flux control at four steps: glucose import, hexokinase, phosphofructokinase, and lactate export (and not at any steps of lower glycolysis). The four flux-controlling steps are specifically upregulated by the Ras oncogene: optogenetic Ras activation rapidly induces the transcription of isozymes catalyzing these four steps and enhances glycolysis. At least one isozyme catalyzing each of these four steps is consistently elevated in human tumors. Thus, in the studied contexts, flux control in glycolysis is concentrated in four key enzymatic steps. Upregulation of these steps in tumors likely underlies the Warburg effect.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HEPES -Lösung, 1 M, sterile-filtered, BioReagent
Sigma-Aldrich
Carbenicillin Dinatriumsalz, 89.0-100.5% anhydrous basis
Sigma-Aldrich
Monoklonaler Anti-α-Tubulin-Antikörper in Maus hergestellte Antikörper, clone DM1A, ascites fluid
Sigma-Aldrich
Fetales Kälberserum, USA origin, Dialyzed by ultrafiltration against 0.15 M NaCl, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Picrotoxin, powder
Sigma-Aldrich
SB-505124 hydrochloride hydrate, ≥98% (HPLC)