Merck
  • 7-Hydroxy-benzopyran-4-one derivatives: a novel pharmacophore of peroxisome proliferator-activated receptor alpha and -gamma (PPARalpha and gamma) dual agonists.
7-Hydroxy-benzopyran-4-one derivatives: a novel pharmacophore of peroxisome proliferator-activated receptor alpha and -gamma (PPARalpha and gamma) dual agonists.
Journal of medicinal chemistry (2009-10-08)
Azadeh Matin, Navnath Gavande, Moon S Kim, Nancy X Yang, Noeris K Salam, Jane R Hanrahan, Rebecca H Roubin, David E Hibbs
ZUSAMMENFASSUNG
Design, synthesis, and in vitro bioevaluation of a new class of potential dual PPARalpha and gamma agonists discovered through a structure-driven design paradigm are described. The 7-hydroxy-benzopyran-4-one moiety (includes flavones, flavanones, and isoflavones) is the key pharmacophore of these novel molecules, exhibiting similarity to the core structure of both fibrates and thiazolidinediones. New lead PPAR ligands were identified from "natraceuticals" and synthetic analogues. In total, 77 molecules, including chalcones, flavones, flavanones, isoflavones, and pyrazole derivatives, were screened and structure-activity relationship studies of the dual agonists undertaken. Compounds 68, 70, 72, and 76 were identified as novel and potent dual PPARalpha and gamma agonists. These novel molecules may have the potential to be the future leads in PPAR-related disorders, including type II diabetes mellitus and metabolic syndrome.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Sigma-Aldrich
Daidzein, ≥98%, synthetic
Sigma-Aldrich
Chrysin, ≥96.5%
Sigma-Aldrich
Diosmetin
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)