Merck
  • CTTNBP2 Controls Synaptic Expression of Zinc-Related Autism-Associated Proteins and Regulates Synapse Formation and Autism-like Behaviors.
CTTNBP2 Controls Synaptic Expression of Zinc-Related Autism-Associated Proteins and Regulates Synapse Formation and Autism-like Behaviors.
Cell reports (2020-06-04)
Pu-Yun Shih, Bing-Yuan Hsieh, Ming-Hui Lin, Tzyy-Nan Huang, Ching-Yen Tsai, Wen-Li Pong, Sue-Ping Lee, Yi-Ping Hsueh
ZUSAMMENFASSUNG
Synaptic dysregulation is a critical feature of autism spectrum disorders (ASDs). Among various autism-associated genes, cortactin binding protein 2 (CTTNBP2) is a cytoskeleton regulator predominantly expressed in neurons and highly enriched at dendritic spines. Here, using Cttnbp2 knockout and ASD-linked mutant mice, we demonstrate that Cttnbp2 deficiency reduces zinc levels in the brain, alters synaptic protein targeting, impairs dendritic spine formation and ultrastructure of postsynaptic density, and influences neuronal activation and autism-like behaviors. A link to autism, the NMDAR-SHANK pathway, and zinc-related regulation are three features shared by CTTNBP2-regulated synaptic proteins. Zinc supplementation rescues the synaptic expression of CTTNBP2-regulated proteins. Moreover, zinc supplementation and administration of D-cycloserine, an NMDAR coagonist, improve the social behaviors of Cttnbp2-deficient mice. We suggest that CTTNBP2 controls the synaptic expression of a set of zinc-regulated autism-associated genes and influences NMDAR function and signaling, providing an example of how genetic and environmental factor crosstalk controls social behaviors.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Zinksulfat Heptahydrat, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-Glutamatrezeptor-2-Antikörper, extrazellulär, Klon 6C4, clone 6C4, Chemicon®, from mouse
Sigma-Aldrich
Anti-metabotroper-Glutamatrezeptor-5-Antikörper, Schmerz, Chemicon®, from rabbit
Sigma-Aldrich
Anti-NMDAR1-Antikörper, Klon 54.1, clone 54.1, Chemicon®, from mouse
Sigma-Aldrich
Anti-PSD95-Antikörper, Klon K28/43, clone K28/43, from mouse
Sigma-Aldrich
Anti-GluR1-NT(-NT)-Antikörper, Klon RH95, clone RH95, from mouse
Sigma-Aldrich
Anti-Guinea Pig IgG (whole molecule)−Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Millipore
ReBlot Plus Kit
Sigma-Aldrich
Anti-NR2A-Antikörper, Upstate®, from rabbit
Sigma-Aldrich
Anti-NMDAR2B-Antikörper, Chemicon®, from rabbit
Sigma-Aldrich
Anti-TUC-4-Protein-Antikörper, serum, Chemicon®