- Arginine-pyrimidine conjugates with therapeutic and prophylactic activity in lethal bacterial infections.
Arginine-pyrimidine conjugates with therapeutic and prophylactic activity in lethal bacterial infections.
Bioorganic & medicinal chemistry letters (2009-10-13)
Dieter Haebich, Hein-Peter Kroll, Hans-Georg Lerchen
PMID19819137
ZUSAMMENFASSUNG
Arginine-pyrimidine conjugates represent a novel class of compounds that exhibits therapeutic and prophylactic activity in lethal infections by Gram-positive and Gram-negative bacteria without showing antibacterial activity in vitro.