Merck
Tessaric acid derivatives induce G2/M cell cycle arrest in human solid tumor cell lines.
Bioorganic & medicinal chemistry (2009-08-12)
Leticia G León, Osvaldo J Donadel, Carlos E Tonn, José M Padrón
ZUSAMMENFASSUNG
A series of analogs were synthesized in a straightforward manner from naturally available sesquiterpenes ilicic acid and tessaric acid. The in vitro antiproliferative activities were examined in the human solid tumor cell lines A2780, HBL-100, HeLa, SW1573, T-47D and WiDr. The most potent analog induced considerably growth inhibition in the range 1.9-4.5 microM. Cell cycle studies for tessaric acid derivatives indicated a prominent arrest of the cell cycle at the G(2)/M phase. Damage to the cells was permanent as determine by the so called 24+24 drug schedule.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
(S)-(+)-Camptothecin, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorouracil, meets USP testing specifications