Merck
Hypoxia drives the assembly of the multienzyme purinosome complex.
The Journal of biological chemistry (2020-05-23)
Cyrielle Doigneaux, Anthony M Pedley, Ishna N Mistry, Monika Papayova, Stephen J Benkovic, Ali Tavassoli
ZUSAMMENFASSUNG
The purinosome is a dynamic metabolic complex composed of enzymes responsible for de novo purine biosynthesis, whose formation has been associated with elevated purine demand. However, the physiological conditions that govern purinosome formation in cells remain unknown. Here, we report that purinosome formation is up-regulated in cells in response to a low-oxygen microenvironment (hypoxia). We demonstrate that increased purinosome assembly in hypoxic human cells requires the activation of hypoxia inducible factor 1 (HIF-1) and not HIF-2. Hypoxia-driven purinosome assembly was inhibited in cells lacking 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMP cyclohydrolase (ATIC), a single enzyme in de novo purine biosynthesis, and in cells treated with a small molecule inhibitor of ATIC homodimerization. However, despite the increase in purinosome assembly in hypoxia, we observed no associated increase in de novo purine biosynthesis in cells. Our results indicate that this was likely due to a reduction in mitochondrial one-carbon metabolism, resulting in reduced mitochondrion-derived one-carbon units needed for de novo purine biosynthesis. The findings of our study further clarify and deepen our understanding of purinosome formation by revealing that this process does not solely depend on cellular purine demand.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Deferoxamin -mesylat (Salz), powder, ≥92.5% (TLC)
Sigma-Aldrich
2-Desoxy-D-Glucose, ≥99% (GC), crystalline
Sigma-Aldrich
Monoklonaler Anti-β-Aktin−Peroxidase-Antikörper der Maus in Maus hergestellte Antikörper, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Hypoxanthin, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-Maus-IgG- & -IgM-Antikörper der Ziege, HRP-Konjugat, 0.8 mg/mL, Chemicon®