Merck
  • HGF protected against diabetic nephropathy via autophagy-lysosome pathway in podocyte by modulating PI3K/Akt-GSK3β-TFEB axis.
HGF protected against diabetic nephropathy via autophagy-lysosome pathway in podocyte by modulating PI3K/Akt-GSK3β-TFEB axis.
Cellular signalling (2020-08-23)
Bo Hou, Yankun Li, Xue Li, Congying Zhang, Zhonghua Zhao, Qi Chen, Nong Zhang, Hui Li
ZUSAMMENFASSUNG
Podocyte loss is a detrimental feature and major cause of proteinuria in diabetic nephropathy (DN). Our previous study revealed that hepatocyte growth factor (HGF) prevented high glucose-induced podocyte injury via enhancing autophagy. In the current study, we aimed to assess the role of HGF on podocyte homeostasis in DN and clarify its mechanisms further. Diabetic mice treated with HGF had markedly reduced ratio of kidney weight to body weight, urinary albumin excretion, podocyte loss and matrix expansion compared with that in the non-treated counterpart. Simultaneously, HGF-treated diabetic mice exhibited increased autophagy activity as indicated by the decreased accumulation of sequestosome 1 (SQSTM1/ p62) and increased microtubule-associated proteins 1 light chains 3 (LC3) II/LC3I ratio. These beneficial effects of HGF were blocked by HGF/c-Met inhibitor Crizotinib or phosphatidylinositide 3-kinases (PI3K) inhibitor LY294002. Moreover, HGF treatment obviously prevented inactivation of the protein kinase B (Akt)-glycogen synthase kinase 3 beta (GSK3β)-transcription factor EB (TFEB) axis in high glucose-stimulated podocytes, which was associated with improved lysosome function and autophagy. Accordingly, adenovirus vector encoding constitutively active GSK3β (Ad-GSK3β-S9A) offset whereas small interfering RNA against GSK3β (GSK3β siRNA) recapitulated salutary effects of HGF on lysosome number and autophagy in podocytes. These results suggested that HGF protected against diabetic nephropathy through restoring podocyte autophagy, which at least partially involved PI3K/Akt-GSK3β-TFEB axis-mediated lysosomal function improvement.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Streptozocin, ≥75% α-anomer basis, ≥98% (HPLC), powder
Sigma-Aldrich
LY-294.002 -hydrochlorid, solid, ≥98% (HPLC)
Sigma-Aldrich
Crizotinib, ≥98% (HPLC)