Merck
QSAR study of antioxidant activity of wine polyphenols.
European journal of medicinal chemistry (2008-04-12)
Vesna Rastija, Marica Medić-Sarić
ZUSAMMENFASSUNG
Quantitative structure activity relationships (QSAR) were obtained describing the antioxidant activity of the main pharmacologically active polyphenols of wine, using molecular properties and descriptors derived from the 2D and 3D representations of molecules. The best models for the prediction of the ability to scavenge the ABTS radical cation were obtained by polynomial regression analysis using zero-order connectivity index and molar refractivity. Statistically, significant models for lipid peroxidation inhibiting effects of flavonoids were obtained by polynomial and multiple regression using lipophilicity, Balaban index, Balaban-type index and 3D GETAWAY descriptor. The 3D descriptors possess the ability for discrimination of stereoisomers, like catechin and epicatechin. We demonstrated that a size and shape of molecules, as well as steric properties, play an important role in the antioxidant activity of polyphenols.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Sigma-Aldrich
Gallussäure, 97.5-102.5% (titration)
Sigma-Aldrich
Kaffeesäure, ≥98.0% (HPLC)
Sigma-Aldrich
trans-Ferulasäure, 99%
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Sigma-Aldrich
(−)-Epicatechin, ≥90% (HPLC)
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Sigma-Aldrich
Gallussäure Monohydrat, ACS reagent, ≥98.0%
Sigma-Aldrich
Gallussäure Monohydrat, ≥99% (HPLC)
Sigma-Aldrich
(−)-Epicatechin, ≥98% (HPLC), from green tea
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Sigma-Aldrich
Myricetin, ≥96.0% (HPLC)
Sigma-Aldrich
trans-Ferulasäure, ≥99%
Sigma-Aldrich
Myricetin, ≥96.0%, crystalline
Supelco
Resveratrol, analytical standard
Supelco
trans-Ferulasäure, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Supelco
(−)-Epicatechin, analytical standard
Supelco
Kaffeesäure, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Supelco
trans-Ferulasäure, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland