Merck
eIF4E S209 phosphorylation licenses myc- and stress-driven oncogenesis.
eLife (2020-11-03)
Hang Ruan, Xiangyun Li, Xiang Xu, Brian J Leibowitz, Jingshan Tong, Lujia Chen, Luoquan Ao, Wei Xing, Jianhua Luo, Yanping Yu, Robert E Schoen, Nahum Sonenberg, Xinghua Lu, Lin Zhang, Jian Yu
ZUSAMMENFASSUNG
To better understand a role of eIF4E S209 in oncogenic translation, we generated EIF4ES209A/+ heterozygous knockin (4EKI) HCT 116 human colorectal cancer (CRC) cells. 4EKI had little impact on total eIF4E levels, cap binding or global translation, but markedly reduced HCT 116 cell growth in spheroids and mice, and CRC organoid growth. 4EKI strongly inhibited Myc and ATF4 translation, the integrated stress response (ISR)-dependent glutamine metabolic signature, AKT activation and proliferation in vivo. 4EKI inhibited polyposis in ApcMin/+ mice by suppressing Myc protein and AKT activation. Furthermore, p-eIF4E was highly elevated in CRC precursor lesions in mouse and human. p-eIF4E cooperated with mutant KRAS to promote Myc and ISR-dependent glutamine addiction in various CRC cell lines, characterized by increased cell death, transcriptomic heterogeneity and immune suppression upon deprivation. These findings demonstrate a critical role of eIF4E S209-dependent translation in Myc and stress-driven oncogenesis and as a potential therapeutic vulnerability.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Y-27632 -dihydrochlorid, ≥98% (HPLC)
Sigma-Aldrich
Nicotinamid, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Kristallviolett, certified by the BSC
Sigma-Aldrich
SB 202190, ≥98% (HPLC)
Sigma-Aldrich
[Leu15]-Gastrin I Human, ≥95% (HPLC)
Sigma-Aldrich
N-Acetylcysteinamid, ≥98% (HPLC)