Merck
  • Transcriptome Sequencing and Comparative Analysis of Amphoteric ESCs and PGCs in Chicken (Gallus gallus).
Transcriptome Sequencing and Comparative Analysis of Amphoteric ESCs and PGCs in Chicken (Gallus gallus).
Animals : an open access journal from MDPI (2020-12-03)
Kai Jin, Jing Zhou, Qisheng Zuo, Jiuzhou Song, Yani Zhang, Guobing Chang, Guohong Chen, Bichun Li
ZUSAMMENFASSUNG
Chicken (Gallus gallus) pluripotent embryonic stem cells (ESCs) and primordial germ cells (PGCs) can be broadly applied in the research of developmental and embryonic biology, but the difference between amphoteric ESCs and PGCs is still elusive. This study determined the sex of collected samples by identifying specific sex markers via polymerase chain reaction (PCR) and fluorescence activated cell sorter (FACS). RNA-seq was utilized to investigate the transcriptomic profile of amphoteric ESCs and PGCs in chicken. The results showed no significant differentially expressed genes (DEGs) in amphoteric ESCs and 227 DEGs exhibited in amphoteric PGCs. Moreover, those 227 DEGs were mainly enriched in 17 gene ontology (GO) terms and 27 pathways according to Kyoto Encyclopedia of Genes and Genomes (KEGG) analysis. Furthermore, qRT-PCR was performed to verify RNA-seq results, and the results demonstrated that Notch1 was highly expressed in male PGCs. In summary, our results provided a knowledge base of chicken amphoteric ESCs and PGCs, which is helpful for future research in relevant biological processes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
MEM Nicht-essentielle Aminosäure-Lösung (100×), without L-glutamine, liquid, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
hBFGF, FGF-Basic, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
ESGRO® Rekombinantes Maus-LIF-Protein, ESGRO Leukemia Inhibitory Factor (LIF) supplement for mouse ES cell culture. Each vial contains 10^6 units/ml