Merck
Lipid peroxidation regulates long-range wound detection through 5-lipoxygenase in zebrafish.
Nature cell biology (2020-09-02)
Anushka Katikaneni, Mark Jelcic, Gary F Gerlach, Yanan Ma, Michael Overholtzer, Philipp Niethammer
ZUSAMMENFASSUNG
Rapid wound detection by distant leukocytes is essential for antimicrobial defence and post-infection survival1. The reactive oxygen species hydrogen peroxide and the polyunsaturated fatty acid arachidonic acid are among the earliest known mediators of this process2-4. It is unknown whether or how these highly conserved cues collaborate to achieve wound detection over distances of several hundreds of micrometres within a few minutes. To investigate this, we locally applied arachidonic acid and skin-permeable peroxide by micropipette perfusion to unwounded zebrafish tail fins. As in wounds, arachidonic acid rapidly attracted leukocytes through dual oxidase (Duox) and 5-lipoxygenase (Alox5a). Peroxide promoted chemotaxis to arachidonic acid without being chemotactic on its own. Intravital biosensor imaging showed that wound peroxide and arachidonic acid converged on half-millimetre-long lipid peroxidation gradients that promoted leukocyte attraction. Our data suggest that lipid peroxidation functions as a spatial redox relay that enables long-range detection of early wound cues by immune cells, outlining a beneficial role for this otherwise toxic process.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
α-Linoleic acid, ≥98%
Sigma-Aldrich
Linolensäure, ≥99%
Sigma-Aldrich
Oleinsäure, ≥99% (GC)
Sigma-Aldrich
α-Tocopherol, ≥95.5%
Sigma-Aldrich
Ethyl-3-aminobenzoat -methansulfonat, 98%
Sigma-Aldrich
Sudanschwarz B, certified by the BSC
Sigma-Aldrich
Diphenyleniodoniumchlorid, ≥98%
Sigma-Aldrich
Arachidonsäure, from non-animal source, ≥98.5% (GC)
Supelco
Ethyl-3-Aminobenzoat -methansulfonat (Salz), analytical standard
Sigma-Aldrich
Liproxstatin-1, >98% (HPLC)
Sigma-Aldrich
Ebselen, cysteine modifier
Sigma-Aldrich
Arachinsäure, synthetic, ≥99.0% (GC)