Merck
  • Increased GABAergic development in iPSC-derived neurons from patients with sporadic Alzheimer's disease.
Increased GABAergic development in iPSC-derived neurons from patients with sporadic Alzheimer's disease.
Neuroscience letters (2020-07-03)
Yueyu Tang, Yingying Han, Hongxiang Yu, Bei Zhang, Gang Li
ZUSAMMENFASSUNG
Alzheimer's disease (AD) is the most common cause of dementia in the elderly, and the underlying molecular mechanisms of this neurodegenerative disorder are still unclear. γ-Aminobutyric acid (GABA) neurons play an essential role in the excitatory/inhibitory (E/I) balance in the brain, and the GABAergic system may contribute to the pathogenesis of AD. We used human induced pluripotent stem cells (iPSCs) generated from sporadic AD (SAD) patients to analyze the phenotype and transcriptional profiles of SAD iPSC-derived neural cells. We observed reduced neurogenesis and increased astrogenesis in SAD neural differentiation. We discovered elevated levels of GABA, glutamate decarboxylase 67 (GAD67), and vesicular GABA transporter (vGAT) in SAD neurons that indicated increased GABAergic development. Gene expression profiling of SAD neural cultures showed upregulation of the GABAergic signaling pathway and downregulation of the neurogenesis pathway. We presumed that the GABAergic transmission system might be enhanced in SAD neurons, as an early pathological change of SAD, which provides a novel target and new direction for the development of more effective therapeutic strategies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Laminin aus Engelbreth-Holm-Swarm murine sarcoma basement membrane, 1-2 mg/mL in Tris-buffered saline, 0.2 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Poly-L-ornithin -Lösung, mol wt 30,000-70,000, 0.01%, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Y-27632 -dihydrochlorid, ≥98% (HPLC)
Sigma-Aldrich
4′,6-Diamidin-2-phenylindol -dihydrochlorid, powder, BioReagent, suitable for cell culture, ≥98% (HPLC), suitable for fluorescence