Merck
  • Synthesis and biological evaluation of nitrogen-containing chalcones as possible anti-inflammatory and antioxidant agents.
Synthesis and biological evaluation of nitrogen-containing chalcones as possible anti-inflammatory and antioxidant agents.
Bioorganic & medicinal chemistry letters (2009-12-17)
Babasaheb P Bandgar, Sachin A Patil, Rajesh N Gacche, Balaji L Korbad, Balwant S Hote, Santosh N Kinkar, Shivkumar S Jalde
ZUSAMMENFASSUNG
A novel series of nitrogen-containing chalcones were synthesized by Mannich reaction and were screened for anti-inflammatory related activities such as inhibition of cyclooxygenase-2 (COX-2), trypsin and beta-glucuronidase. The antioxidant potential was demonstrated using 1,1-diphenyl-2-picryl hydrazine (DPPH) radical scavenging activity. The results of the above studies shows that the compounds synthesized were found to be effective inhibitors of above pro-inflammatory enzymes, and were found to be possess moderate radical scavenging potential. Overall, the results of the studies reveal that the chalcones with N-methyl piperazine methyl and piperidine methyl substitution (4c, 3b, 4d, 6b) seems to be important for inhibition of beta-glucuronidase. Whereas the chalcones with piperidine methyl substitution (8b, 7b, 7c, 6c, 4b, 3c, 3b) were observed as effective inhibitors of COX-2, while the same compounds were found to be less reactive against COX-1 as compared to COX-2.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salicylsäure, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetylsalicylsäure, ≥99.0%
Sigma-Aldrich
Aspirin, meets USP testing specifications
Sigma-Aldrich
Salicylsäure, BioXtra, ≥99.0%
Sigma-Aldrich
Salicylsäure, ≥99%, FG
Sigma-Aldrich
Salicylsäure, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Acetylsalicylsäure, analytical standard
Sigma-Aldrich
Salicylsäure, suitable for plant cell culture
Sigma-Aldrich
Salicylsäure, ReagentPlus®, ≥99%
Sigma-Aldrich
Salicylsäure, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Supelco
Salicylsäure, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland