Merck
Pharmacology of terazosin: an alpha 1-selective blocker.
Journal of clinical pharmacology (1993-09-01)
J J Kyncl
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Terazosin hydrochloride, ≥98% (TLC), powder