Merck
  • Bioaugmentation with copper tolerant endophyte Pseudomonas lurida strain EOO26 for improved plant growth and copper phytoremediation by Helianthus annuus.
Bioaugmentation with copper tolerant endophyte Pseudomonas lurida strain EOO26 for improved plant growth and copper phytoremediation by Helianthus annuus.
Chemosphere (2020-12-05)
Adarsh Kumar, Tripti, Olga Voropaeva, Maria Maleva, Ksenia Panikovskaya, Galina Borisova, Mani Rajkumar, L Benedict Bruno
ZUSAMMENFASSUNG
Organic fertilizers became a better alternative to chemical fertilizers in modern agricultural practices however, contamination of copper (Cu) from organic fertilizer is still a major concern for the globe. Plant growth promoting (PGP) microorganisms showed their efficiency to combat with this problem and thus Cu tolerant PGP endophytes from roots of Odontarrhena obovata (Alyssum obovatum) growing on Cu smelter contaminated serpentine soil were explored in present study. Out of twenty-four isolates, Pseudomonas lurida strain EOO26 identified by 16s rRNA gene sequencing was selected to check its efficacy for Cu-remediation. The strain EOO26 showed multi-metal tolerance, drought resistance and exhibited PGP attributes such as 1-aminocyclopropane-1-carboxylic acid (ACC) deaminase, siderophore and ammonia production. Significant production of indole-3-acetic acid and phosphate-solubilization under different Cu concentration (0-100 mg L-1) at varying pH (5.0-8.0) suggests potentiality of this strain to work effectively under wide range of abiotic stress conditions. Plant growth experiment (pH 6.8 ± 0.3) in copper spiked soil suggested a significant increase in length and dry weight of root and shoot of sunflower (Helianthus annuus) after inoculation with strain EOO26. Plants inoculated with strain EOO26 resulted in increase in Cu uptake by 8.6-fold for roots and 1.9-fold for leaves than uninoculated plants. The total plant uptake in inoculated Cu treatment was 2.6-fold higher than uninoculated one, which is much higher than the previously reported Cu accumulating plants. The excellent adaptation abilities and promising metal removal efficiency strongly indicate superiority of strain EOO26 for phytoremediation of Cu-contamination and may work effectively for Cu removal from contaminated soils.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Indol-3-essigsäure Natriumsalz, suitable for plant cell culture, BioReagent, ≥98%