Merck
  • Short-term Cudrania tricuspidata fruit vinegar administration attenuates obesity in high-fat diet-fed mice by improving fat accumulation and metabolic parameters.
Short-term Cudrania tricuspidata fruit vinegar administration attenuates obesity in high-fat diet-fed mice by improving fat accumulation and metabolic parameters.
Scientific reports (2020-12-05)
Jun-Hui Choi, Myung-Kon Kim, Soo-Hwan Yeo, Seung Kim
ZUSAMMENFASSUNG
Previous studies have suggested that vinegar intake can help to reduce body fat and hyperglycemia. Therefore, this study aimed to evaluate the anti-obesity efficacy of vinegar fermented using Cudrania tricuspidata fruits (CTFV) and its main phenolic constituents and to analyze its molecular mechanism and changes in obesity-related metabolizing enzymatic activities. We found that HFD significantly caused hepatic steatosis; increases in body fats, feed efficiency, liver mass, lipids, insulin, oxidative parameters, cardiovascular-associated risk indices, lipase and α-amylase activities, whereas CTFV efficaciously attenuated HFD-induced oxidant stress, fat accumulation, obesity-related enzymatic activity, and the activation or reduction of obesity-related molecular reactions via improving metabolic parameters including phosphorylated insulin receptor substrate 1, protein tyrosine phosphatase 1B, phosphorylated phosphoinositide 3-kinase/protein kinase B, phosphorylated mitogen-activated protein kinases, sterol regulatory element-binding protein 1c, CCAAT/enhancer-binding protein, and fatty acid synthase; and decreases in adiponectin receptor 1, leptin receptor, adenosine monophosphate-activated protein kinase, acetyl-CoA carboxylase, and peroxisome proliferator-activated receptor, subsequently ameliorating HFD-induced obesity. Therefore, CTFV might provide a functional food resource or nutraceutical product for reducing body fat accumulation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Millipore
Ratten-/Maus-Insulin-ELISA, This Rat/Mouse Insulin ELISA is used to measure & quantify Insulin levels in Metabolism & Endocrine research.
Sigma-Aldrich
Buttersäure-4-Nitrophenylester, ≥98%
Rutin, phyproof® Reference Substance
Millipore
Maus-Leptin-ELISA, This Mouse Leptin ELISA is used to measure & quantify Leptin levels in Metabolism & Endocrine & Neuroscience research.
Millipore
Mouse Adiponectin ELISA, This Mouse Adiponectin ELISA is used to measure & quantify Adiponectin levels in Metabolism & Endocrine research.
Sigma-Aldrich
4-Nitrophenyl phenylphosphonate, adenosine receptor agonist, 5′-Nucleotide Phosphodiesterase substrate