Merck
Cell Types Promoting Goosebumps Form a Niche to Regulate Hair Follicle Stem Cells.
Cell (2020-07-18)
Yulia Shwartz, Meryem Gonzalez-Celeiro, Chih-Lung Chen, H Amalia Pasolli, Shu-Hsien Sheu, Sabrina Mai-Yi Fan, Farnaz Shamsi, Steven Assaad, Edrick Tai-Yu Lin, Bing Zhang, Pai-Chi Tsai, Megan He, Yu-Hua Tseng, Sung-Jan Lin, Ya-Chieh Hsu
ZUSAMMENFASSUNG
Piloerection (goosebumps) requires concerted actions of the hair follicle, the arrector pili muscle (APM), and the sympathetic nerve, providing a model to study interactions across epithelium, mesenchyme, and nerves. Here, we show that APMs and sympathetic nerves form a dual-component niche to modulate hair follicle stem cell (HFSC) activity. Sympathetic nerves form synapse-like structures with HFSCs and regulate HFSCs through norepinephrine, whereas APMs maintain sympathetic innervation to HFSCs. Without norepinephrine signaling, HFSCs enter deep quiescence by down-regulating the cell cycle and metabolism while up-regulating quiescence regulators Foxp1 and Fgf18. During development, HFSC progeny secretes Sonic Hedgehog (SHH) to direct the formation of this APM-sympathetic nerve niche, which in turn controls hair follicle regeneration in adults. Our results reveal a reciprocal interdependence between a regenerative tissue and its niche at different stages and demonstrate sympathetic nerves can modulate stem cells through synapse-like connections and neurotransmitters to couple tissue production with demands.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
4-Hydroxytamoxifen, ≥70% Z isomer (remainder primarily E-isomer)
Sigma-Aldrich
Anti-Tyrosin-Hydroxylase-Antikörper, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Sox9-Antikörper, Chemicon®, from rabbit
Sigma-Aldrich
Monoklonaler Anti-Aktin, α-Glattmuskel - Cy3 Maus-Antikörper, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Collagenase aus Clostridium histolyticum, lyophilized powder, ≥125 CDU/mg solid (CDU = collagen digestion units), 0.5-5.0 FALGPA units/mg solid
Sigma-Aldrich
Corticosteron, ≥98.5% (HPLC)
Sigma-Aldrich
6-Hydroxydopamin -hydrobromid, 95%
Sigma-Aldrich
Doxycyclin -hydrochlorid
Sigma-Aldrich
Anti-Tyrosin-Hydroxylase-Antikörper, Chemicon®, from sheep
Sigma-Aldrich
DL-Isoproterenol -hydrochlorid
Sigma-Aldrich
Anti-β-Tubulin-III in Kaninchen hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Procaterol hydrochloride