Merck
  • Therapeutic activity of an inhaled potent SARS-CoV-2 neutralizing human monoclonal antibody in hamsters.
Therapeutic activity of an inhaled potent SARS-CoV-2 neutralizing human monoclonal antibody in hamsters.
Cell reports. Medicine (2021-03-03)
Michael S Piepenbrink, Jun-Gyu Park, Fatai S Oladunni, Ashlesha Deshpande, Madhubanti Basu, Sanghita Sarkar, Andreas Loos, Jennifer Woo, Phillip Lovalenti, Derek Sloan, Chengjin Ye, Kevin Chiem, Christopher W Bates, Reuben E Burch, Nathaniel B Erdmann, Paul A Goepfert, Vu L Truong, Mark R Walter, Luis Martinez-Sobrido, James J Kobie
ZUSAMMENFASSUNG
SARS-CoV-2 infection results in viral burden in the respiratory tract, enabling transmission and leading to substantial lung pathology. The 1212C2 fully human monoclonal antibody was derived from an IgM memory B cell of a COVID-19 patient, has high affinity for the Spike protein receptor binding domain, neutralizes SARS-CoV-2, and exhibits in vivo prophylactic and therapeutic activity in hamsters when delivered intraperitoneally, reducing upper and lower respiratory viral burden and lung pathology. Inhalation of nebulized 1212C2 at levels as low as 0.6 mg/kg, corresponding to 0.03 mg/kg lung-deposited dose, reduced the viral burden below the detection limit and mitigated lung pathology. The therapeutic efficacy of an exceedingly low dose of inhaled 1212C2 supports the rationale for local lung delivery for dose-sparing benefits, as compared to the conventional parenteral route of administration. These results suggest that the clinical development of 1212C2 formulated and delivered via inhalation for the treatment of SARS-CoV-2 infection should be considered.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Formalinlösung, neutral gepuffert, 10 %, histological tissue fixative
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Supelco
Avicel® PH-101, ~50 μm particle size
Sigma-Aldrich
Anti-SARS-CoV-1/2 NP Antibody, clone 1C7C7 ZooMAb® Mouse Monoclonal, recombinant, expressed in HEK 293 cells