Merck
  • Inhibition of SARS-CoV-2 by Highly Potent Broad-Spectrum Anti-Coronaviral Tylophorine-Based Derivatives.
Inhibition of SARS-CoV-2 by Highly Potent Broad-Spectrum Anti-Coronaviral Tylophorine-Based Derivatives.
Frontiers in pharmacology (2021-02-02)
Cheng-Wei Yang, Yue-Zhi Lee, Hsing-Yu Hsu, Jia-Tsrong Jan, Yi-Ling Lin, Sui-Yuan Chang, Tzu-Ting Peng, Ruey-Bing Yang, Jian-Jong Liang, Chun-Che Liao, Tai-Ling Chao, Yu-Hau Pang, Han-Chieh Kao, Wen-Zheng Huang, Jiunn-Horng Lin, Chun-Ping Chang, Guang-Hao Niu, Szu-Huei Wu, Huey-Kang Sytwu, Chiung-Tong Chen, Shiow-Ju Lee
ZUSAMMENFASSUNG
Tylophorine-based compounds and natural cardiotonic steroids (cardenolides and bufadienolides) are two classes of transmissible gastroenteritis coronavirus inhibitors, targeting viral RNA and host cell factors, respectively. We tested both types of compounds against two types of coronaviruses, to compare and contrast their antiviral properties, and with view to their further therapeutic development. Examples of both types of compounds potently inhibited the replication of both feline infectious peritonitis virus and human coronavirus OC43 with EC50 values of up to 8 and 16 nM, respectively. Strikingly, the tylophorine-based compounds tested inhibited viral yields of HCoV-OC43 to a much greater extent (7-8 log magnitudes of p.f.u./ml) than the cardiotonic steroids (about 2-3 log magnitudes of p.f.u./ml), as determined by end point assays. Based on these results, three tylophorine-based compounds were further examined for their anti-viral activities on two other human coronaviruses, HCoV-229E and SARS-CoV-2. These three tylophorine-based compounds inhibited HCoV-229E with EC50 values of up to 6.5 nM, inhibited viral yields of HCoV-229E by 6-7 log magnitudes of p.f.u./ml, and were also found to inhibit SARS-CoV-2 with EC50 values of up to 2.5-14 nM. In conclusion, tylophorine-based compounds are potent, broad-spectrum inhibitors of coronaviruses including SARS-CoV-2, and could be used for the treatment of COVID-19.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cell Counting Kit - 8, for quantitation of viable cell number in proliferation and cytotoxicity assays
Sigma-Aldrich
Kristallviolett, certified by the BSC
Sigma-Aldrich
Ouabain Octahydrat, ≥95% (HPLC), powder
Sigma-Aldrich
Digitoxin, ≥92% (HPLC), powder
Supelco
Digoxin, analytical standard
Sigma-Aldrich
Anti-Coronavirus Group Antigen Antibody, nucleoprotein of OC-43, clone 542-7D, clone 542-7D, Chemicon®, from mouse